Learn More
Description
Specifications
Specifications
| Acidity | 0.2 mL |
| CAS Max % | 1 |
| Boiling Point | 189 to 190°C |
| Packaging | Amber Glass Bottle |
| Refractive Index | 1.500-1.503 |
| Quantity | 100 mL |
| Specific Gravity | 1.011-1.016 |
| Percent Purity | 95 to 105% |
| Grade | USP |
| Physical Form | Liquid |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
