Learn More
Cayman Chemical Octanoic Acid-13C, Purity: ≥98%, CAS: 59669-16-8, 250 mg
SDP
Catalog No. 502992346
Encompass_Preferred
An internal standard for the quantification of octanoic acid by GC- or LC-MS
Catalog No. 50-299-2346 Supplier Cayman Chemical Supplier No. 29461250 MG
An internal standard for the quantification of octanoic acid by GC- or LC-MS