Learn More
Cayman Chemical Octanoic Acid-d15, Purity: ≥99% deuterated forms (d1-d15), CAS: 69974-55-6, 100 mg
SDP
Catalog No. 502937506
Encompass_Preferred
An internal standard for the quantification of octanoic acid by GC- or LC-MS
Catalog No. 50-293-7506 Supplier Cayman Chemical Supplier No. 29074100 MG
An internal standard for the quantification of octanoic acid by GC- or LC-MS