Learn More
Cayman Chemical Octanoic Acid-d2, Purity: ≥99% deuterated forms (d1-d2); ≤1% d0, CAS: 64118-36-1, 1 mg
SDP
Catalog No. 502988659
Encompass_Preferred
An internal standard for the quantification of octanoic acid by GC- or LC-MS
Catalog No. 50-298-8659 Supplier Cayman Chemical Supplier No. 294121 MG
An internal standard for the quantification of octanoic acid by GC- or LC-MS