Learn More
Cayman Chemical Octanoic Acid-d3, Purity: ≥99% deuterated forms (d1-d3), CAS: 156779-05-4, 10 mg
SDP
Catalog No. 502937029
Encompass_Preferred
An internal standard for the quantification of octanoic acid by GC- or LC-MS
Catalog No. 50-293-7029 Supplier Cayman Chemical Supplier No. 2886910 MG
An internal standard for the quantification of octanoic acid by GC- or LC-MS