Learn More
Cayman Chemical OctnoIc AcId-d3 25mg
SDP
Catalog No. 502937404
Encompass_Preferred
An internal standard for the quantification of octanoic acid by GC- or LC-MS
Catalog No. 50-293-7404 Supplier Cayman Chemical Supplier No. 2886925 MG
An internal standard for the quantification of octanoic acid by GC- or LC-MS