Learn More
Cayman Chemical Octanoic Acid-d3, Purity: ≥99% deuterated forms (d1-d3), CAS: 156779-05-4, 50 mg
SDP
Catalog No. 502940354
Encompass_Preferred
An internal standard for the quantification of octanoic acid by GC- or LC-MS
Catalog No. 50-294-0354 Supplier Cayman Chemical Supplier No. 2886950 MG
An internal standard for the quantification of octanoic acid by GC- or LC-MS