Learn More
Cayman Chemical Octanoic Acid methyl ester, Purity: ≥98%, CAS: 111-11-5, 500 g
SDP
Catalog No. 502936931
Encompass_Preferred
A fatty acid methyl ester; has been found in biodiesels made from the transesterification of beef tallow, soybean oil, and babassu oil blends; an aromatic volatile compound in cantaloupe, galia, and honeydew melons
Catalog No. 50-293-6931 Supplier Cayman Chemical Supplier No. 26717500 G
A fatty acid methyl ester; has been found in biodiesels made from the transesterification of beef tallow, soybean oil, and babassu oil blends; an aromatic volatile compound in cantaloupe, galia, and honeydew melons