Learn More
Cayman Chemical Octanoyl-Coenzyme A (sodium salt), Purity: ≥95%, 1 mg
SDP
Catalog No. 502940981
Encompass_Preferred
A medium-chain acyl CoA; levels increased in liver in patients with Reye’s syndrome and β-oxidation of octanoyl-CoA by MCADH is decreased in patients with MCD; inhibits citrate synthase and glutamate dehydrogenase
Catalog No. 50-294-0981 Supplier Cayman Chemical Supplier No. 278681 MG
A medium-chain acyl CoA; levels increased in liver in patients with Reye’s syndrome and β-oxidation of octanoyl-CoA by MCADH is decreased in patients with MCD; inhibits citrate synthase and glutamate dehydrogenase