Learn More
Cayman Chemical Octnoyl-Coenzyme AsodIum s 5mg
SDP
Catalog No. 502940235
Encompass_Preferred
A medium-chain acyl CoA; levels increased in liver in patients with Reye’s syndrome and β-oxidation of octanoyl-CoA by MCADH is decreased in patients with MCD; inhibits citrate synthase and glutamate dehydrogenase
Catalog No. 50-294-0235 Supplier Cayman Chemical Supplier No. 278685 MG
A medium-chain acyl CoA; levels increased in liver in patients with Reye’s syndrome and β-oxidation of octanoyl-CoA by MCADH is decreased in patients with MCD; inhibits citrate synthase and glutamate dehydrogenase