Learn More
Cayman Chemical Octanoyl-DL-carnitine (chloride), Purity: ≥98%, CAS: 14919-35-8, 50 mg
SDP
Catalog No. 502972862
Encompass_Preferred
A medium-chain acylcarnitine; decreases the oxidation rate of 3-methyl-2-butanoate and 4-methyl-2-oxopentanoate in isolated rat muscle mitochondria in the presence of carnitine, but increases it in the absence of carnitine; with malate, has been used as a mitochondrial respiration substrate along with malate to measure the effect of creatine on the respiration rate of isolated rat cardiac fibers exposed to increasing concentrations of ADP
Catalog No. 50-297-2862 Supplier Cayman Chemical Supplier No. 1504850 MG
A medium-chain acylcarnitine; decreases the oxidation rate of 3-methyl-2-butanoate and 4-methyl-2-oxopentanoate in isolated rat muscle mitochondria in the presence of carnitine, but increases it in the absence of carnitine; with malate, has been used as a mitochondrial respiration substrate along with malate to measure the effect of creatine on the respiration rate of isolated rat cardiac fibers exposed to increasing concentrations of ADP