Learn More
Cayman Chemical Octanoyl-L-carnitine (chloride), Purity: ≥95%, CAS: 54377-02-5, 10 mg
SDP
Catalog No. 502934834
Encompass_Preferred
A medium-chain acylcarnitine; the physiologically active form of octanoyl-DL-carnitine; plasma levels are elevated in patients with end-stage renal disease on continuous ambulatory PD compared with both patients on automated PD and healthy individuals; does not undergo hydrolysis in the blood or during sample preparation when used as a standard for the quantification of octanoylcarnitine
Catalog No. 50-293-4834 Supplier Cayman Chemical Supplier No. 2655810 MG
A medium-chain acylcarnitine; the physiologically active form of octanoyl-DL-carnitine; plasma levels are elevated in patients with end-stage renal disease on continuous ambulatory PD compared with both patients on automated PD and healthy individuals; does not undergo hydrolysis in the blood or during sample preparation when used as a standard for the quantification of octanoylcarnitine