Learn More
Cayman Chemical OctnoylCoenzyme AsodIum s 10mg
SDP
Catalog No. 502940223
Encompass_Preferred
A medium-chain acyl CoA; levels increased in liver in patients with Reye’s syndrome and β-oxidation of octanoyl-CoA by MCADH is decreased in patients with MCD; inhibits citrate synthase and glutamate dehydrogenase
Catalog No. 50-294-0223 Supplier Cayman Chemical Supplier No. 2786810 MG
A medium-chain acyl CoA; levels increased in liver in patients with Reye’s syndrome and β-oxidation of octanoyl-CoA by MCADH is decreased in patients with MCD; inhibits citrate synthase and glutamate dehydrogenase