Learn More
Medchemexpress LLC Octocrylene-d10 | 2714435-95-5 | 371.54 | C24H17D10NO2 | 1mg
SDP
Catalog No. 5000270422
Encompass_Preferred
Octocrylene-d10 is the deuterium labeled Octocrylene Octocrylene is an organic ultraviolet (UV) filter which absorbs mainly UVB radiation and short UVA wavelengths[1]
Catalog No. 50-002-70422 Supplier Medchemexpress LLC Supplier No. HYW7499661MG
Octocrylene-d10 is the deuterium labeled Octocrylene Octocrylene is an organic ultraviolet (UV) filter which absorbs mainly UVB radiation and short UVA wavelengths[1]