Learn More
Medchemexpress LLC Octodrine hydrochloride | 5984-59-8 | 98.0% | 1 ML
SDP
Catalog No. 5000412926
Encompass_Preferred
Octodrine hydrochloride is a central nervous activator that functions by increasing the uptake of dopamine and noradrenaline. It has been observed to increase pain threshold, cardiac rate, and myocardial contractility.
  • Increases dopamine and noradrenaline uptake
  • Increases pain threshold
  • Positive chronotropic effect on cardiac rate
  • Positive inotropic effect on myocardial contractility
  • For research use only
Catalog No. 50-004-12926 Supplier Medchemexpress LLC Supplier No. HYB1233A10MM/1ML
Octodrine hydrochloride is a central nervous activator that functions by increasing the uptake of dopamine and noradrenaline. It has been observed to increase pain threshold, cardiac rate, and myocardial contractility.
  • Increases dopamine and noradrenaline uptake
  • Increases pain threshold
  • Positive chronotropic effect on cardiac rate
  • Positive inotropic effect on myocardial contractility
  • For research use only