Learn More
Medchemexpress LLC Octopamine-d3 | 111988-19-3 | 99.5% | 1 MG
SDP
Catalog No. 5000407569
Encompass_Preferred
Octopamine-d3 is a deuterium labeled form of Octopamine, intended for research use only. This white to off-white solid can be used as a tracer or as an internal standard for quantitative analysis.
  • Deuterium labeled Octopamine
  • Used as a tracer
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Deuteration can impact pharmacokinetic and metabolic profiles of drugs
  • Appears as a white to off-white solid
Catalog No. 50-004-07569 Supplier Medchemexpress LLC Supplier No. HYB0528S1MG
Octopamine-d3 is a deuterium labeled form of Octopamine, intended for research use only. This white to off-white solid can be used as a tracer or as an internal standard for quantitative analysis.
  • Deuterium labeled Octopamine
  • Used as a tracer
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Deuteration can impact pharmacokinetic and metabolic profiles of drugs
  • Appears as a white to off-white solid