Learn More
Medchemexpress LLC Octopamine-d4 hydrochloride | 1219803-62-9 | 98.0% | 1 MG
SDP
Catalog No. 5000407010
Encompass_Preferred
Octopamine-d4 hydrochloride is a deuterium-labeled form of Octopamine hydrochloride. It acts as a neurohormone, neuromodulator, and neurotransmitter in invertebrates, stimulating alpha2-adrenoceptors. It has been observed to increase glycogenolysis, glycolysis, oxygen uptake, gluconeogenesis, and portal perfusion pressure.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-004-07010 Supplier Medchemexpress LLC Supplier No. HYB0528AS1MG
Octopamine-d4 hydrochloride is a deuterium-labeled form of Octopamine hydrochloride. It acts as a neurohormone, neuromodulator, and neurotransmitter in invertebrates, stimulating alpha2-adrenoceptors. It has been observed to increase glycogenolysis, glycolysis, oxygen uptake, gluconeogenesis, and portal perfusion pressure.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS