Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Octopamine-d4 hydrochloride | 1219803-62-9 | 98.3% | 5 MG
SDP

Catalog No. 5000407612
Encompass_Preferred

Octopamine-d4 (hydrochloride) is the deuterium labeled Octopamine hydrochloride. Octopamine ((±)-p-Octopamine) hydrochloride, a biogenic monoamine structurally related to noradrenaline, acts as a neurohormone, a neuromodulator and a neurotransmitter in invertebrates. Octopamine hydrochloride can stimulate alpha2-adrenoceptors (ARs) in Chinese hamster ovary cells transfected with human alpha2-ARs. It has been shown to increase glycogenolysis, glycolysis, oxygen uptake, gluconeogenesis and the portal perfusion pressure.

  • Molecular formula: C8H9D3CINO2
  • Molecular weight: 192.66
  • Target: Adrenergic receptor; endogenous metabolite
  • Pathway: GPCR/G protein; neuronal signaling; metabolic enzyme/protease
  • Storage: 4°C, sealed storage, away from moisture. In solvent: -80°C for 6 months; -20°C for 1 month (sealed storage, away from moisture).
  • Solubility (in vitro): DMSO: ≥ 100 mg/mL (519.05 mM); H2O: ≥ 50 mg/mL (259.52 mM)

Catalog No. 50-004-07612 Supplier Medchemexpress LLC Supplier No. HYB0528AS5MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Octopamine-d4 (hydrochloride) is the deuterium labeled Octopamine hydrochloride. Octopamine ((±)-p-Octopamine) hydrochloride, a biogenic monoamine structurally related to noradrenaline, acts as a neurohormone, a neuromodulator and a neurotransmitter in invertebrates. Octopamine hydrochloride can stimulate alpha2-adrenoceptors (ARs) in Chinese hamster ovary cells transfected with human alpha2-ARs. It has been shown to increase glycogenolysis, glycolysis, oxygen uptake, gluconeogenesis and the portal perfusion pressure.

  • Molecular formula: C8H9D3CINO2
  • Molecular weight: 192.66
  • Target: Adrenergic receptor; endogenous metabolite
  • Pathway: GPCR/G protein; neuronal signaling; metabolic enzyme/protease
  • Storage: 4°C, sealed storage, away from moisture. In solvent: -80°C for 6 months; -20°C for 1 month (sealed storage, away from moisture).
  • Solubility (in vitro): DMSO: ≥ 100 mg/mL (519.05 mM); H2O: ≥ 50 mg/mL (259.52 mM)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.