Learn More
Medchemexpress LLC Octopamine-d4 hydrochloride | 1219803-62-9 | MFCD11983520 | 98.3% | 10 MG
SDP
Catalog No. 5000297363
Encompass_Preferred
Octopamine-d4 hydrochloride is the deuterium-labeled (d4) hydrochloride salt of octopamine supplied as a solid stable-isotope standard for analytical and research use. It is intended for use as an internal standard in mass spectrometry, tracer studies, and quantitative analyses where isotopic labeling improves accuracy.
  • Deuterium-labeled internal standard for mass spectrometry and quantitative analysis.
  • High purity (~98.3%) suitable for analytical applications.
  • Solid, light brown to brown appearance for easy handling and storage.
  • Recommended storage: sealed at 4°C; in solvent, -80°C up to 6 months.
  • Available in small mg pack sizes for analytical workflows.
Catalog No. 50-002-97363 Supplier Medchemexpress LLC Supplier No. HYB0528AS10MG
Octopamine-d4 hydrochloride is the deuterium-labeled (d4) hydrochloride salt of octopamine supplied as a solid stable-isotope standard for analytical and research use. It is intended for use as an internal standard in mass spectrometry, tracer studies, and quantitative analyses where isotopic labeling improves accuracy.
  • Deuterium-labeled internal standard for mass spectrometry and quantitative analysis.
  • High purity (~98.3%) suitable for analytical applications.
  • Solid, light brown to brown appearance for easy handling and storage.
  • Recommended storage: sealed at 4°C; in solvent, -80°C up to 6 months.
  • Available in small mg pack sizes for analytical workflows.