Learn More
Apexbio Technology LLC (+,-)-Octopamine HCl 770-05-8 10mM (in 1mL DMSO)
SDP
Catalog No. 5000093053
Encompass_Preferred
( -)-Octopamine HCl (CAS 770-05-8) is a biogenic monoamine structurally analogous to noradrenaline In invertebrates it functions as a neurohormone neuromodulator and neurotransmitter by interacting with specific octopamine receptors to regulate physiological and behavioral processes including metabolism locomotion and learning The compound is widely utilized in neurobiological research to investigate aminergic signaling pathways synaptic transmission and the functional roles of monoamines in invertebrate nervous systems Its use facilitates studies on neurotransmitter mechanisms distinct from those predominant in vertebrates
Catalog No. 50-000-93053 Supplier Apexbio Technology LLC Supplier No. B19955.1
( -)-Octopamine HCl (CAS 770-05-8) is a biogenic monoamine structurally analogous to noradrenaline In invertebrates it functions as a neurohormone neuromodulator and neurotransmitter by interacting with specific octopamine receptors to regulate physiological and behavioral processes including metabolism locomotion and learning The compound is widely utilized in neurobiological research to investigate aminergic signaling pathways synaptic transmission and the functional roles of monoamines in invertebrate nervous systems Its use facilitates studies on neurotransmitter mechanisms distinct from those predominant in vertebrates