Learn More
Apexbio Technology LLC (+,-)-Octopamine HCl 770-05-8 1g
SDP
Catalog No. 5000092883
Encompass_Preferred
( -)-Octopamine HCl (CAS 770-05-8) is a biogenic monoamine structurally analogous to noradrenaline In invertebrates it functions as a neurohormone neuromodulator and neurotransmitter by interacting with specific octopamine receptors to regulate physiological and behavioral processes including metabolism locomotion and learning The compound is widely utilized in neurobiological research to investigate aminergic signaling pathways synaptic transmission and the functional roles of monoamines in invertebrate nervous systems Its use facilitates studies on neurotransmitter mechanisms distinct from those predominant in vertebrates
Catalog No. 50-000-92883 Supplier Apexbio Technology LLC Supplier No. B19951000
Add to Cart
Add to Cart
( -)-Octopamine HCl (CAS 770-05-8) is a biogenic monoamine structurally analogous to noradrenaline In invertebrates it functions as a neurohormone neuromodulator and neurotransmitter by interacting with specific octopamine receptors to regulate physiological and behavioral processes including metabolism locomotion and learning The compound is widely utilized in neurobiological research to investigate aminergic signaling pathways synaptic transmission and the functional roles of monoamines in invertebrate nervous systems Its use facilitates studies on neurotransmitter mechanisms distinct from those predominant in vertebrates