Learn More
Medchemexpress LLC Octopamine hydrochloride | 770-05-8 | 99.9% | 1 G
SDP
Catalog No. 5000405459
Encompass_Preferred
Octopamine hydrochloride is a biogenic monoamine structurally related to noradrenaline. It functions as a neurohormone, neuromodulator, and neurotransmitter in invertebrates, stimulating alpha2-adrenoceptors and increasing various metabolic processes. It mediates behaviors like learning and memory, regulates endocrine gland activity, and mobilizes lipids and carbohydrates through specific G protein-coupled receptors.
  • Acts as a neurohormone, neuromodulator, and neurotransmitter
  • Stimulates alpha2-adrenoceptors (ARs)
  • Increases glycogenolysis, glycolysis, oxygen uptake, gluconeogenesis, and portal perfusion pressure
  • Regulates sensory input desensitization, arousal, and rhythmic behaviors
  • Involved in complex behaviors like learning and memory
  • Regulates endocrine gland activity
  • Induces lipid and carbohydrate mobilization
Catalog No. 50-004-05459 Supplier Medchemexpress LLC Supplier No. HYB0528A1G
Octopamine hydrochloride is a biogenic monoamine structurally related to noradrenaline. It functions as a neurohormone, neuromodulator, and neurotransmitter in invertebrates, stimulating alpha2-adrenoceptors and increasing various metabolic processes. It mediates behaviors like learning and memory, regulates endocrine gland activity, and mobilizes lipids and carbohydrates through specific G protein-coupled receptors.
  • Acts as a neurohormone, neuromodulator, and neurotransmitter
  • Stimulates alpha2-adrenoceptors (ARs)
  • Increases glycogenolysis, glycolysis, oxygen uptake, gluconeogenesis, and portal perfusion pressure
  • Regulates sensory input desensitization, arousal, and rhythmic behaviors
  • Involved in complex behaviors like learning and memory
  • Regulates endocrine gland activity
  • Induces lipid and carbohydrate mobilization