Learn More
Medchemexpress LLC Octopamine hydrochloride | 770-05-8 | 99.9% | 1 ML
SDP
Catalog No. 5000407572
Encompass_Preferred
Octopamine ((±)-p-Octopamine) hydrochloride, a biogenic monoamine structurally related to noradrenaline, acts as a neurohormone, a neuromodulator and a neurotransmitter in invertebrates. It can stimulate alpha2-adrenoceptors (ARs) in Chinese hamster ovary cells transfected with human alpha2-ARs. Research indicates Octopamine hydrochloride increases glycogenolysis, glycolysis, oxygen uptake, gluconeogenesis and the portal perfusion pressure.
  • Acts as a neurohormone, neuromodulator, and neurotransmitter in invertebrates.
  • Stimulates alpha2-adrenoceptors (ARs).
  • Increases glycogenolysis and glycolysis.
  • Enhances oxygen uptake.
  • Boosts gluconeogenesis.
  • Elevates portal perfusion pressure.
Catalog No. 50-004-07572 Supplier Medchemexpress LLC Supplier No. HYB0528A10MM/1ML
Octopamine ((±)-p-Octopamine) hydrochloride, a biogenic monoamine structurally related to noradrenaline, acts as a neurohormone, a neuromodulator and a neurotransmitter in invertebrates. It can stimulate alpha2-adrenoceptors (ARs) in Chinese hamster ovary cells transfected with human alpha2-ARs. Research indicates Octopamine hydrochloride increases glycogenolysis, glycolysis, oxygen uptake, gluconeogenesis and the portal perfusion pressure.
  • Acts as a neurohormone, neuromodulator, and neurotransmitter in invertebrates.
  • Stimulates alpha2-adrenoceptors (ARs).
  • Increases glycogenolysis and glycolysis.
  • Enhances oxygen uptake.
  • Boosts gluconeogenesis.
  • Elevates portal perfusion pressure.