Learn More
Medchemexpress LLC Octopamine hydrochloride | 770-05-8 | 99.9% | 25 MG
SDP
Catalog No. 5000450483
Encompass_Preferred
Octopamine hydrochloride is a biogenic monoamine structurally related to noradrenaline. It functions as a neurohormone, neuromodulator, and neurotransmitter in invertebrates. This analytical standard is intended for research and analytical applications.
  • Stimulates alpha2-adrenoceptors (ARs) in Chinese hamster ovary cells
  • Increases glycogenolysis, glycolysis, oxygen uptake, gluconeogenesis, and portal perfusion pressure
  • Serves as an analytical standard
  • Suitable for research and analytical applications
Catalog No. 50-004-50483 Supplier Medchemexpress LLC Supplier No. 25MGHYB0528AR
Octopamine hydrochloride is a biogenic monoamine structurally related to noradrenaline. It functions as a neurohormone, neuromodulator, and neurotransmitter in invertebrates. This analytical standard is intended for research and analytical applications.
  • Stimulates alpha2-adrenoceptors (ARs) in Chinese hamster ovary cells
  • Increases glycogenolysis, glycolysis, oxygen uptake, gluconeogenesis, and portal perfusion pressure
  • Serves as an analytical standard
  • Suitable for research and analytical applications