Learn More
Medchemexpress LLC Octopamine hydrochloride | 770-05-8 | 99.9% | 250 MG
SDP
Catalog No. 5000405722
Encompass_Preferred
Octopamine hydrochloride is a biogenic monoamine structurally related to noradrenaline, acting as a neurohormone, neuromodulator, and neurotransmitter in invertebrates. It can stimulate alpha2-adrenoceptors (ARs) and is observed to increase glycogenolysis, glycolysis, oxygen uptake, gluconeogenesis, and portal perfusion pressure.
  • Acts as a neurohormone, neuromodulator, and neurotransmitter
  • Stimulates alpha2-adrenoceptors (ARs)
  • Regulates desensitization of sensory inputs and arousal
  • Involved in initiation and maintenance of rhythmic behaviors
  • Regulates endocrine gland activity
  • Induces mobilization of lipids and carbohydrates
Catalog No. 50-004-05722 Supplier Medchemexpress LLC Supplier No. 250MGHYB0528AR
Octopamine hydrochloride is a biogenic monoamine structurally related to noradrenaline, acting as a neurohormone, neuromodulator, and neurotransmitter in invertebrates. It can stimulate alpha2-adrenoceptors (ARs) and is observed to increase glycogenolysis, glycolysis, oxygen uptake, gluconeogenesis, and portal perfusion pressure.
  • Acts as a neurohormone, neuromodulator, and neurotransmitter
  • Stimulates alpha2-adrenoceptors (ARs)
  • Regulates desensitization of sensory inputs and arousal
  • Involved in initiation and maintenance of rhythmic behaviors
  • Regulates endocrine gland activity
  • Induces mobilization of lipids and carbohydrates