Learn More
Medchemexpress LLC Octopamine hydrochloride | 770-05-8 | MFCD00012881 | 99.90% | 5 G
SDP
Catalog No. 5000407963
Encompass_Preferred
Octopamine hydrochloride is a biogenic monoamine structurally related to noradrenaline. It functions as a neurohormone, neuromodulator, and neurotransmitter in invertebrates, stimulating alpha2-adrenoceptors. This compound is known to affect various physiological processes, including metabolism and neuroregulation. It achieves its effects by binding to specific G protein-coupled receptors.
  • Regulates sensory input desensitization, arousal, and complex behaviors like learning and memory.
  • Modulates endocrine gland activity.
  • Mobilizes lipids and carbohydrates as a neurohormone.
  • Stimulates alpha2-adrenoceptors.
  • Increases glycogenolysis, glycolysis, oxygen uptake, gluconeogenesis, and portal perfusion pressure.
Catalog No. 50-004-07963 Supplier Medchemexpress LLC Supplier No. HYB0528A5G
Octopamine hydrochloride is a biogenic monoamine structurally related to noradrenaline. It functions as a neurohormone, neuromodulator, and neurotransmitter in invertebrates, stimulating alpha2-adrenoceptors. This compound is known to affect various physiological processes, including metabolism and neuroregulation. It achieves its effects by binding to specific G protein-coupled receptors.
  • Regulates sensory input desensitization, arousal, and complex behaviors like learning and memory.
  • Modulates endocrine gland activity.
  • Mobilizes lipids and carbohydrates as a neurohormone.
  • Stimulates alpha2-adrenoceptors.
  • Increases glycogenolysis, glycolysis, oxygen uptake, gluconeogenesis, and portal perfusion pressure.