Learn More
Medchemexpress LLC Octopamine hydrochloride (Standard) | 770-05-8 | 99.9%
SDP
Catalog No. 5000408844
Encompass_Preferred
Octopamine hydrochloride (Standard) is an analytical standard intended for research and analytical applications. This biogenic monoamine acts as a neurohormone, neuromodulator, and neurotransmitter in invertebrates. It stimulates alpha2-adrenoceptors (ARs) in Chinese hamster ovary cells and has been shown to increase glycogenolysis, glycolysis, oxygen uptake, gluconeogenesis, and portal perfusion pressure.
  • Analytical standard for research and analytical applications
  • Functions as a neurohormone, neuromodulator, and neurotransmitter in invertebrates
  • Stimulates alpha2-adrenoceptors (ARs)
  • Increases glycogenolysis, glycolysis, oxygen uptake, gluconeogenesis, and portal perfusion pressure
  • Used in HPLC, GC, and MS research experiments
Catalog No. 50-004-08844 Supplier Medchemexpress LLC Supplier No. HYB0528AR100MG
Octopamine hydrochloride (Standard) is an analytical standard intended for research and analytical applications. This biogenic monoamine acts as a neurohormone, neuromodulator, and neurotransmitter in invertebrates. It stimulates alpha2-adrenoceptors (ARs) in Chinese hamster ovary cells and has been shown to increase glycogenolysis, glycolysis, oxygen uptake, gluconeogenesis, and portal perfusion pressure.
  • Analytical standard for research and analytical applications
  • Functions as a neurohormone, neuromodulator, and neurotransmitter in invertebrates
  • Stimulates alpha2-adrenoceptors (ARs)
  • Increases glycogenolysis, glycolysis, oxygen uptake, gluconeogenesis, and portal perfusion pressure
  • Used in HPLC, GC, and MS research experiments