Learn More
Abcam 8-Octanoylo x ypyrene-1,3,6-trisulfonic acid trisodium salt, 25MG
Catalog No. 50996243
Encompass_Preferred
MW 650.6 Da. Highly polar and water soluble lipase substrate derived from HPTS (pyranine), which forms tight micelles in aqueous solutions. Lipase activity can be established by measuring the fluorescence kinetics following the increase in emission of HPTS (hydroxypyrene trisulfonic acid). Due to its low pKa value (7.3), it is partially dissociated to form an anion that shows strong absorption (max: 460 nm) and maximum intensity at 520 nm. Fluorescence intensity is pH dependent between Ex:440-480 nm and reaches the isosbestic point at Ex: 415 nm.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-996-243 Supplier Abcam Supplier No. AB27532525MG
MW 650.6 Da. Highly polar and water soluble lipase substrate derived from HPTS (pyranine), which forms tight micelles in aqueous solutions. Lipase activity can be established by measuring the fluorescence kinetics following the increase in emission of HPTS (hydroxypyrene trisulfonic acid). Due to its low pKa value (7.3), it is partially dissociated to form an anion that shows strong absorption (max: 460 nm) and maximum intensity at 520 nm. Fluorescence intensity is pH dependent between Ex:440-480 nm and reaches the isosbestic point at Ex: 415 nm.
The product is subject to the following: Abcam Restricted Use Statement