Learn More
Medchemexpress LLC HY-P0036 10mg , Octreotide CAS:83150-76-9 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYP003610MG
Encompass_Preferred
HY-P0036 10mg Octreotide CAS: 83150-76-9 Octreotide (SMS 201-995) is a somatostatin receptor agonist and synthetic octapeptide endogenous somatostatin analogue. Octreotide (SMS 201-995) can bind to the somatostatin receptor and mainly subtypes 2, 3, and 5, increases Gi activity, and reduces intracellular cAMP production. Octreotide (SMS 201-995) has antitumor activity, mediates apoptosis and may also be used in disease studies in acromegaly. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-0320