Learn More
TARGETMOL CHEMICALS INC OCTREOTIDE ACETATE 100MG
SDP
Catalog No. 502576807
Encompass_Preferred
Also available in 5 mg 10 mg 25 mg 50 mg 200 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Octreotide Acetate (Sandostatin) is a potent long-acting synthetic somatostatin octapeptide analog that inhibits secretion of growth hormone. purity: 99%
Catalog No. 50-257-6807 Supplier TARGETMOL CHEMICALS INC Supplier No. T4119100MG
Also available in 5 mg 10 mg 25 mg 50 mg 200 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Octreotide Acetate (Sandostatin) is a potent long-acting synthetic somatostatin octapeptide analog that inhibits secretion of growth hormone. purity: 99%