Learn More
Abcam Octreotide, Somatostatin analog, 1MG
Catalog No. 502706800
Encompass_Preferred
MW 1019.24 g/mol, Purity >98%. Somatostatin analog. Acts as an agonist at somatostatin sst2, sst3 and sst5 receptors (IC₅₀ values are 0.4-2.1, 4.4-34.5 and 5.6-32 nM, respectively). Displays antitumor and antiproliferative effects.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-6800 Supplier Abcam Supplier No. AB1412081MG
MW 1019.24 g/mol, Purity >98%. Somatostatin analog. Acts as an agonist at somatostatin sst2, sst3 and sst5 receptors (IC₅₀ values are 0.4-2.1, 4.4-34.5 and 5.6-32 nM, respectively). Displays antitumor and antiproliferative effects.
The product is subject to the following: Abcam Restricted Use Statement