Learn More
ILEX LIFE SCIENCES LLC OCTREOTIDE SYNTHETIC 1 MG
SDP
Catalog No. 502734007
Encompass_Preferred
Ilex Life Sciences Octreotide Acetate, Synthetic Peptide Hormone (1 mg). DESCRIPTION: Octreotide is a single, non-glycosylated, synthetic polypeptide chain. It contains 8 amino acids and has a molecular mass of 1019.26 Dalton. Octreotide is purified by proprietary chromatographic techniques. CAS REGISTRY NO.: 83150-76-9. MOLECULAR WEIGHT: 1019.26 Dalton. MOLECULAR FORMULA: C49H66N10O10S2. AMINO ACID SEQUENCE: H-D-Phe-Cys-Phe-D-Trp-Lys-Thr-Cys-L-threoninol. PURITY: Purity greater than 98.0% as determined by RP-HPLC. FORMULATION: Sterile-filtered white powder lyophilized from a concentrated (1 mg/ml) solution with no additives. USAGE: This product is intended for Laboratory Research Use Only. Not for use in diagnostic or therapeutic procedures.
Catalog No. 50-273-4007 Supplier ILEX LIFE SCIENCES LLC Supplier No. A226501
Add to Cart
Add to Cart
Ilex Life Sciences Octreotide Acetate, Synthetic Peptide Hormone (1 mg). DESCRIPTION: Octreotide is a single, non-glycosylated, synthetic polypeptide chain. It contains 8 amino acids and has a molecular mass of 1019.26 Dalton. Octreotide is purified by proprietary chromatographic techniques. CAS REGISTRY NO.: 83150-76-9. MOLECULAR WEIGHT: 1019.26 Dalton. MOLECULAR FORMULA: C49H66N10O10S2. AMINO ACID SEQUENCE: H-D-Phe-Cys-Phe-D-Trp-Lys-Thr-Cys-L-threoninol. PURITY: Purity greater than 98.0% as determined by RP-HPLC. FORMULATION: Sterile-filtered white powder lyophilized from a concentrated (1 mg/ml) solution with no additives. USAGE: This product is intended for Laboratory Research Use Only. Not for use in diagnostic or therapeutic procedures.