Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

YMC America, Inc OCTYL 10UM 120A 250X10MM
SDP

Catalog No. 5090005634
Encompass_Preferred

Octyl 10Um; 120A; 250 X 10mm

Catalog No. 50-900-05634 Supplier YMC America, Inc Supplier No. OC12S112510WT
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Octyl 10Um; 120A; 250 X 10mm

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.