Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

YMC America, Inc OCTYL 15UM 200A 250X30MM
SDP

Catalog No. 5090005685
Encompass

Octyl 15Um; 200A 250 X 30mm

Catalog No. 50-900-05685 Supplier YMC America, Inc Supplier No. OC20S162530WT
May include imposed supplier surcharges.
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

Octyl 15Um; 200A 250 X 30mm

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.