Learn More
YMC America, Inc OCTYL 5UM 200A 50X3MM
SDP
Catalog No. 50157119
Encompass_Preferred
OC20S05-0503WT: Octyl, 5um, 200A, 50X3mm Column
Catalog No. 50-157-119 Supplier YMC America, Inc Supplier No. OC20S050503WT
OC20S05-0503WT: Octyl, 5um, 200A, 50X3mm Column