Learn More
Medchemexpress LLC Octyl-a-ketoglutarat 5mg | 876150-14-0 | 5 MG
SDP
Catalog No. 5000229762
Encompass_Preferred
Octyl-α-ketoglutarate is a cell-permeable ester of α-ketoglutarate used in biochemical and cellular research to modulate HIF signaling and prolyl hydroxylase activity. It is used to probe metabolism, demethylation, and TCA cycle dysfunction in cell-based assays.
  • Cell-permeable ester of α-ketoglutarate suitable for cell-based assays.
  • Stimulates prolyl hydroxylase (PHD) activity and modulates HIF signaling.
  • Useful for studies of metabolism, demethylation, and TCA cycle dysfunction.
  • Available in small milligram quantities for research use.
  • Provided with quality documentation and testing.
Catalog No. 50-002-29762 Supplier Medchemexpress LLC Supplier No. HY1143335MG
Octyl-α-ketoglutarate is a cell-permeable ester of α-ketoglutarate used in biochemical and cellular research to modulate HIF signaling and prolyl hydroxylase activity. It is used to probe metabolism, demethylation, and TCA cycle dysfunction in cell-based assays.
  • Cell-permeable ester of α-ketoglutarate suitable for cell-based assays.
  • Stimulates prolyl hydroxylase (PHD) activity and modulates HIF signaling.
  • Useful for studies of metabolism, demethylation, and TCA cycle dysfunction.
  • Available in small milligram quantities for research use.
  • Provided with quality documentation and testing.