Learn More
Cayman Chemical Octyl-a-ketoglutarate 10mg
SDP
Catalog No. 502954661
Encompass_Preferred
A stable, cell-permeable form of α-ketoglutarate which accumulates rapidly and preferentially in cells with a dysfunctional TCA cycle; stimulates PHD activity and increases HIF-1α turnover when used at 1 mM; competitively blocks succinate- or fumarate-mediated inhibition of PHD
Catalog No. 50-295-4661 Supplier Cayman Chemical Supplier No. 1197010 MG
A stable, cell-permeable form of α-ketoglutarate which accumulates rapidly and preferentially in cells with a dysfunctional TCA cycle; stimulates PHD activity and increases HIF-1α turnover when used at 1 mM; competitively blocks succinate- or fumarate-mediated inhibition of PHD