Learn More
Sigma Aldrich Fine Chemicals Biosciences Octyl acetate 98 fCC fG1KG
Catalog No. 502899818
Encompass
Octyl acetate is one of the major constituents of the essential oils of Heracleum crenatifolium oranges and grapefruit.
Catalog No. 50-289-9818 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. W2806071KGK
Octyl acetate is one of the major constituents of the essential oils of Heracleum crenatifolium oranges and grapefruit.