Learn More
Abcam Octyl-beta-D-thioglucopyranoside, APG surfactant
Catalog No. NC3740255
Encompass_Preferred
MW 308.44 Da, Purity >98%. APG surfactant. Reagent for the solubilisation and isolation of membranes.
Catalog No. NC3740255 Supplier Abcam Supplier No. AB141435 SAME LOT
Add to Cart
Add to Cart
Request a Quote
MW 308.44 Da, Purity >98%. APG surfactant. Reagent for the solubilisation and isolation of membranes.