Learn More
World Precision Instrument GLUture Topical Tissue Adhesive
SDP
Catalog No. NC9992277
Encompass
Gluture topical tissue adhesive is an Octyl/Butyl cyanoacrylate blend that offers a flexible seal and a strong bond that contours the wound site. Gluture applies purple for excellent visibility and dries clear. Offered in a multi-use package of 10 applicators designed for quick, flow-controlled application.
Catalog No. NC9992277 Supplier World Precision Instrument Supplier No. 503763
Gluture topical tissue adhesive is an Octyl/Butyl cyanoacrylate blend that offers a flexible seal and a strong bond that contours the wound site. Gluture applies purple for excellent visibility and dries clear. Offered in a multi-use package of 10 applicators designed for quick, flow-controlled application.