Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Octyl β-D-maltopyranoside | 82494-08-4 | MFCD00065526 | 454.51 g/mol | C20H38O11 | 100 MG
SDP

Catalog No. 5000209539
Encompass_Preferred

Octyl β-D-maltopyranoside is a non-ionic surfactant used as a biochemical assay reagent and to solubilize membrane proteins for biochemical and biophysical studies.

  • Non-ionic detergent for membrane protein solubilization
  • Water-soluble, suitable for biochemical assays
  • Molecular weight 454.51 g/mol
  • CAS number 82494-08-4
  • Typical pack sizes listed: 1 MG, 5 MG, 10 MG (manufacturer datasheet)

Catalog No. 50-002-09539 Supplier Medchemexpress LLC Supplier No. HYW040202100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Octyl β-D-maltopyranoside is a non-ionic surfactant used as a biochemical assay reagent and to solubilize membrane proteins for biochemical and biophysical studies.

  • Non-ionic detergent for membrane protein solubilization
  • Water-soluble, suitable for biochemical assays
  • Molecular weight 454.51 g/mol
  • CAS number 82494-08-4
  • Typical pack sizes listed: 1 MG, 5 MG, 10 MG (manufacturer datasheet)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.