Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Octyl β-D-maltopyranoside | 82494-08-4 | MFCD00065526 | >=99.0% | 454.51 g/mol | C20H38O11 | 500 MG
SDP

Catalog No. 5000208923
Encompass_Preferred

Octyl β-D-maltopyranoside is a water-soluble, nonionic detergent and biochemical assay reagent commonly used to solubilize and isolate membrane proteins for biochemical and structural studies.

  • Water-soluble, nonionic detergent suitable for membrane protein solubilization and biochemical assays.
  • Molecular formula C20H38O11; molecular weight ≈ 454.5 g/mol.
  • CAS number 82494-08-4 for unambiguous identification.
  • High purity where specified (≥99.0% by HPLC reported by some suppliers).
  • Available in milligram-scale quantities (e.g., 500 MG pack variants).

Catalog No. 50-002-08923 Supplier Medchemexpress LLC Supplier No. HYW040202500MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Octyl β-D-maltopyranoside is a water-soluble, nonionic detergent and biochemical assay reagent commonly used to solubilize and isolate membrane proteins for biochemical and structural studies.

  • Water-soluble, nonionic detergent suitable for membrane protein solubilization and biochemical assays.
  • Molecular formula C20H38O11; molecular weight ≈ 454.5 g/mol.
  • CAS number 82494-08-4 for unambiguous identification.
  • High purity where specified (≥99.0% by HPLC reported by some suppliers).
  • Available in milligram-scale quantities (e.g., 500 MG pack variants).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.