Learn More
Medchemexpress LLC Octyl gallate | 1034-01-1 | MFCD00002197 | 282.33 | 25 MG
SDP
Catalog No. 5000389741
Encompass_Preferred
Octyl gallate, also known as Progallin O, is a derivative of gallic acid recognized for its diverse biological activities. It is frequently employed as a food additive due to its potent antioxidant and antimicrobial properties. Research indicates its promising effectiveness against a broad spectrum of microorganisms, including both Gram-positive and Gram-negative bacteria. Furthermore, it has demonstrated significant antiviral effects against viruses such as HSV-1, vesicular stomatitis virus (VSV), and poliovirus, as well as notable antifungal activity.
  • Acts as a powerful antioxidant.
  • Exhibits broad-spectrum antibacterial activity.
  • Shows selective antiviral properties.
  • Possesses antifungal effects.
  • Commonly used as a food additive.
Catalog No. 50-003-89741 Supplier Medchemexpress LLC Supplier No. 25MGHYN2011R
Octyl gallate, also known as Progallin O, is a derivative of gallic acid recognized for its diverse biological activities. It is frequently employed as a food additive due to its potent antioxidant and antimicrobial properties. Research indicates its promising effectiveness against a broad spectrum of microorganisms, including both Gram-positive and Gram-negative bacteria. Furthermore, it has demonstrated significant antiviral effects against viruses such as HSV-1, vesicular stomatitis virus (VSV), and poliovirus, as well as notable antifungal activity.
  • Acts as a powerful antioxidant.
  • Exhibits broad-spectrum antibacterial activity.
  • Shows selective antiviral properties.
  • Possesses antifungal effects.
  • Commonly used as a food additive.