Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Octyl isocyanate, 99%

Catalog No. p-177051
Change view
Click to view available options
Quantity:
5 g
10 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC416460050 5 g Glass bottle
AC416460100 10 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC416460050 Supplier Thermo Scientific Chemicals Supplier No. 416460050
Only null left
Add to Cart

CAS: 3158-26-7 | C9H17NO | 155.24 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3158-26-7
Molecular Formula C9H17NO
Molecular Weight (g/mol) 155.24
MDL Number MFCD00013887
InChI Key DYQFCTCUULUMTQ-UHFFFAOYSA-N
Synonym octyl isocyanate, octylisocyanate, octane, 1-isocyanato, n-octylisocyanate, 1-octyl isocyanate, n-octyl isocyanate, 1-isocyanato-octane, isocyanic acid, octyl ester, octanisocyanate
PubChem CID 76616
IUPAC Name 1-isocyanatooctane
SMILES CCCCCCCCN=C=O

Specifications

CAS Min % 98.5
CAS Max % 100.0
Color Yellow
Boiling Point 200°C to 204°C
Flash Point 71°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.4290 to 1.431
Linear Formula CH3(CH2)7NCO
Quantity 5 g
Solubility Information Solubility in water: decomposes in water
Formula Weight 155.24
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Octyl isocyanate, 99%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Harmful in contact with skin.
Causes skin irritation.
May cause respiratory irritation.
Harmful if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Harmful if swallowed.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF INHALED: If breathing is difficult, remove to fresh air and keep at rest in a position comfortable for breathing.
If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 221-598-1
TSCA TSCA

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.