Learn More
Bachem n-Octylpolyoxyethylene
SDP
Supplier:  Bachem 4031122.0025
Encompass_Preferred
n-Octylpolyoxyethylene, 27252-75-1, , Mr , 25g. Octyl-POE, C8En (n = 2 to 9), Rosenbusch-Tenside. Octyl-POE solubilizes membrane proteins. The CMC value is 6.6 mM.
Catalog No. 50-194-7413