Learn More
Medchemexpress LLC Octyl salicylate-d4 | 2469274-09-5 | 99.9% | 254.36 | C15H18D4O3 | 1mg
SDP
Catalog No. 5000270480
Encompass_Preferred
Octisalate-d4 (Octyl salicylate-d4 2-Ethylhexyl salicylate-d4) is the deuterium labeled Octisalate (HY-B0929) Octisalate (Octyl salicylate) is an organic compound with high lipophilicity which is used in sunscreens and cosmetics that absorbs UVB rays[1][2][3]
Catalog No. 50-002-70480 Supplier Medchemexpress LLC Supplier No. HYW7147671MG
Octisalate-d4 (Octyl salicylate-d4 2-Ethylhexyl salicylate-d4) is the deuterium labeled Octisalate (HY-B0929) Octisalate (Octyl salicylate) is an organic compound with high lipophilicity which is used in sunscreens and cosmetics that absorbs UVB rays[1][2][3]