Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Octylamine Hydrochloride 99%, Thermo Scientific™

Catalog No. AC416431000
Change view
Click to view available options
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
AC416431000 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC416431000 Supplier Thermo Scientific Chemicals Supplier No. 416431000

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 142-95-0
CAS Min % 98.5
CAS Max % 100.0
Infrared Spectrum Authentic
Assay Percent Range 99%
Molecular Formula C8H20ClN
Linear Formula CH3(CH2)7NH2·HCl
Quantity 100g
MDL Number MFCD00042019
Synonym octylammonium chloride, octylamine hydrochloride, 1-octanamine, hydrochloride, octylamine, hydrochloride, octylamine-monohydrochloride, 1-octanamine, hydrochloride 1:1, octan-1-aminium chloride, octylamine, chloride, n-octylammonium chloride, n-octylamine hydrochloride
InChI Key PHFDTSRDEZEOHG-UHFFFAOYSA-N
SMILES [H+].[Cl-].CCCCCCCCN
IUPAC Name hydrogen octan-1-amine chloride
Molecular Weight (g/mol) 165.71
PubChem CID 67346
Formula Weight 165.71
Percent Purity ≥98.5%
Chemical Name or Material Octylamine hydrochloride, 99%
Show More Show Less
EINECSNumber 205-574-8
TSCA TSCA
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.