Learn More
Medchemexpress LLC Octylamine (n-octylamine) | 111-86-4 | MFCD00008247 | 99.7% | 129.24 g/mol | C8H19N | 250 G
SDP
Catalog No. 5000302705
Encompass_Preferred
Octylamine (n-octylamine, CAS 111-86-4) is a primary aliphatic amine commonly used as a chemical building block and reagent in organic synthesis, analytical chemistry, and formulation work. It combines a reactive primary amine with a hydrophobic eight-carbon chain, making it useful for derivatization, surfactant research, and other laboratory applications. Handle according to SDS and laboratory safety practices.
  • High purity suitable for synthesis and analytical use.
  • Primary amine functionality for derivatization and coupling reactions.
  • Hydrophobic C8 chain useful in surfactant and formulation studies.
  • Suitable for use in common organic solvents and reaction conditions.
  • Provided in lab-scale pack sizes for research and development workflows.
Catalog No. 50-003-02705 Supplier Medchemexpress LLC Supplier No. 250GHYW032014
Octylamine (n-octylamine, CAS 111-86-4) is a primary aliphatic amine commonly used as a chemical building block and reagent in organic synthesis, analytical chemistry, and formulation work. It combines a reactive primary amine with a hydrophobic eight-carbon chain, making it useful for derivatization, surfactant research, and other laboratory applications. Handle according to SDS and laboratory safety practices.
  • High purity suitable for synthesis and analytical use.
  • Primary amine functionality for derivatization and coupling reactions.
  • Hydrophobic C8 chain useful in surfactant and formulation studies.
  • Suitable for use in common organic solvents and reaction conditions.
  • Provided in lab-scale pack sizes for research and development workflows.