Learn More
Medchemexpress LLC Octylphenol ethoxylate | 9036-19-5 | 99.5% | Average 537 g/mol | t-Oct-C6H4-(OCH2CH2)xOH (x ≈ 7-8) | 50 ML
SDP
Catalog No. 5000282693
Encompass_Preferred
Polyoxyethylene octylphenyl ether is a nonionic ethoxylated surfactant used as a detergent and solubilizing agent in biochemical and industrial processes. It exhibits a low cloud point that enables temperature-dependent phase separation for selective extraction and purification, and is commonly used for membrane solubilization, protein extraction, and endotoxin removal.
  • nonionic surfactant with a low cloud point for reversible phase separation.
  • enables efficient membrane solubilization and protein extraction.
  • facilitates endotoxin removal and phase-partition purification workflows.
  • liquid form allows easy dispensing and mixing into aqueous solutions.
  • average molecular weight ~537 g/mol, suitable for biochemical applications.
Catalog No. 50-002-82693 Supplier Medchemexpress LLC Supplier No. HYW13442250ML
Polyoxyethylene octylphenyl ether is a nonionic ethoxylated surfactant used as a detergent and solubilizing agent in biochemical and industrial processes. It exhibits a low cloud point that enables temperature-dependent phase separation for selective extraction and purification, and is commonly used for membrane solubilization, protein extraction, and endotoxin removal.
  • nonionic surfactant with a low cloud point for reversible phase separation.
  • enables efficient membrane solubilization and protein extraction.
  • facilitates endotoxin removal and phase-partition purification workflows.
  • liquid form allows easy dispensing and mixing into aqueous solutions.
  • average molecular weight ~537 g/mol, suitable for biochemical applications.