Learn More
Carbosynth LLC. OD1 Toxin, 1mg, Disulfide bonds: C13-C64, C17-C37, C23-C47, and C27-C49, MW: 7206.21, Molecular formula: C308H466N90O95S8
SDP
Catalog No. 501686070
Encompass_Preferred
Synonyms: Asp-Lys-Asn-Cys-Val-Tyr-Thr-Cys-Ala-Ser-Asn-Gly-Tyr-Cys-Asn-Thr-Glu-Cys-Thr-Lys-Asn-Gly-Ala-Glu-Ser-Gly-Tyr-Cys-Gln-Trp-Ile-Gly-Arg-Tyr-Gly-Asn-Ala-Cys-Trp-Cys-Ile-Lys-Leu-Pro-Asp-Glu-Val-Pro-Ile-Arg-Ile-Pro-Gly-Lys-Cys-Arg-NH2, Application: A Potent Nav1.7 Channel Activator Originally Isolated from the venom of the scorpion, Odonthobuthus doriae, Storage Temperature: -20°C, References: A. Jalali, et al., FEBS Lett., 579, 4181 (2005)., T. Durek, et al., Chem. Biol., 8, 1215 (2013).
Catalog No. 50-168-6070 Supplier Carbosynth LLC. Supplier No. ODT3866PI1MG
Synonyms: Asp-Lys-Asn-Cys-Val-Tyr-Thr-Cys-Ala-Ser-Asn-Gly-Tyr-Cys-Asn-Thr-Glu-Cys-Thr-Lys-Asn-Gly-Ala-Glu-Ser-Gly-Tyr-Cys-Gln-Trp-Ile-Gly-Arg-Tyr-Gly-Asn-Ala-Cys-Trp-Cys-Ile-Lys-Leu-Pro-Asp-Glu-Val-Pro-Ile-Arg-Ile-Pro-Gly-Lys-Cys-Arg-NH2, Application: A Potent Nav1.7 Channel Activator Originally Isolated from the venom of the scorpion, Odonthobuthus doriae, Storage Temperature: -20°C, References: A. Jalali, et al., FEBS Lett., 579, 4181 (2005)., T. Durek, et al., Chem. Biol., 8, 1215 (2013).